Learn More
N-Boc-propargylamine, 97%
Product Code. 15430188
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430188 25 g 25g
15410188 1 g 1g
15420188 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430188 Supplier Thermo Scientific Alfa Aesar Supplier No. H53406.14
Request Bulk or Custom Format
This item is not returnable. View return policy
CAS: 92136-39-5 | C8H13NO2 | 155.197 g/mol
N-Boc-propargylamine is used to prepare triazolobenzylidene-thiazolopyrimidines. which act as CDC25 phosphatase inhibitors. Further, it is used for the synthesis of beta-glucan polysaccharide analogs. In addition to this, it is involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-disubstituted cyclopentenones .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-propargylamine is used to prepare triazolobenzylidene-thiazolopyrimidines. which act as CDC25 phosphatase inhibitors. Further, it is used for the synthesis of beta-glucan polysaccharide analogs. In addition to this, it is involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-disubstituted cyclopentenones .
Solubility
Soluble in chloroform.
Notes
Moisture sensitive. Incompatible with oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 92136-39-5
Molecular Formula C8H13NO2
Molecular Weight (g/mol) 155.197
MDL Number MFCD07367245
InChI Key DSPYCWLYGXGJNJ-UHFFFAOYSA-N
Synonym n-boc-propargylamine, n-tert-butoxycarbonyl propargylamine, n-boc-prop-2-ynylamine, tert-butyl prop-2-yn-1-ylcarbamate, n-boc-propargyl amine, tert-butyl n-prop-2-yn-1-yl carbamate, tert-butyl prop-2-ynylcarbamate, n-boc-2-propyn-1-amine, n-boc propargyl amine, prop-2-ynyl-carbamic acid tert-butyl ester
PubChem CID 11073648
IUPAC Name tert-butyl N-prop-2-ynylcarbamate
SMILES CC(C)(C)OC(=O)NCC#C
Specifications
Melting Point 40°C to 44°C
Linear Formula HC≡CCH2NHCO2C(CH3)3
Solubility Information Soluble in chloroform.
Quantity 25 g
Flash Point 93°C (199°F)
Formula Weight 155.2
Sensitivity Moisture sensitive
Percent Purity 97%
Chemical Name or Material N-Boc-propargylamine