missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-bencil-L-treonina, 99 %, Thermo Scientific Chemicals

Catalog Number p-7025938
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
15260-10-3
C16H23NO5
309.36
MFCD00066062
CTXPLTPDOISPTE-YPMHNXCESA-N
boc-thr bzl-oh, boc-o-benzyl-l-threonine, n-tert-butoxycarbonyl-o-benzyl-l-threonine, n-boc-o-benzyl-l-threonine, boc-thr bzl, o-benzyl-n-tert-butoxycarbonyl-l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-threonine, l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2s,3r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxybutanoic acid
1549483
ácido (2S,3R)-2-[(2-metilpropan-2-il)oxicarbonilamino]-3-fenilmetoxibutanoico
C[C@@H](OCC1=CC=CC=C1)[C@H](NC(=O)OC(C)(C)C)C(O)=O
Este artículo no se puede devolver. Vea la política de devoluciones
15260-10-3
C16H23NO5
309.36
MFCD00066062
CTXPLTPDOISPTE-YPMHNXCESA-N
boc-thr bzl-oh, boc-o-benzyl-l-threonine, n-tert-butoxycarbonyl-o-benzyl-l-threonine, n-boc-o-benzyl-l-threonine, boc-thr bzl, o-benzyl-n-tert-butoxycarbonyl-l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-threonine, l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2s,3r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxybutanoic acid
1549483
ácido (2S,3R)-2-[(2-metilpropan-2-il)oxicarbonilamino]-3-fenilmetoxibutanoico
C[C@@H](OCC1=CC=CC=C1)[C@H](NC(=O)OC(C)(C)C)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-Boc-O-benzyl-L-threonine
Punto de fusión 115°C to 116°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 309.36
Porcentaje de pureza 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'