Learn More
Alfa Aesar™ N-Boc-N-methyl-L-phenylalanine dicyclohexylammonium salt, 97%
Marca:  Alfa Aesar™ H62318.14
Detalles adicionales : CAS : 40163-88-0 Peso : 0.02500kg
Cantidad. 25g
 Ver más versiones de este producto
Código de producto. 15447286
Este producto se ha descatalogado y ya no está en venta. Consulte el producto para saber si se ofrecen alternativas o póngase en contacto con nuestro equipo de Atención al Cliente en el número 902 239 303.
Especificaciones
40163-88-0
460.659
MFORGORUOHHLHW-YDALLXLXSA-N
16211321
CC(C)(C)OC(=O)N(C)C(CC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2
C27H44N2O4
MFCD00070397
boc-n-me-phe dcha, boc-n-me-phe.dcha, z-l-neopentylglycine dicyclohexylammonium salt, boc-mephe-oh.dcha, boc-n-me-phe-ohcha, boc-n-me-l-phe-oh x dcha, boc-n-me-phe-oh dicyclohexylammonium salt, n-boc-n-methyl-l-phenylalanine dicyclohexylammonium salt, boc-n-me-phe-oh dicyclohexylammonium salt chn, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-phenylalanine dicyclohexylamine
N-cyclohexylcyclohexanamine;(2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-phenylpropanoic acid
Especificaciones
N-Boc-N-methyl-L-phenylalanine dicyclohexylammonium salt
C27H44N2O4
boc-n-me-phe dcha, boc-n-me-phe.dcha, z-l-neopentylglycine dicyclohexylammonium salt, boc-mephe-oh.dcha, boc-n-me-phe-ohcha, boc-n-me-l-phe-oh x dcha, boc-n-me-phe-oh dicyclohexylammonium salt, n-boc-n-methyl-l-phenylalanine dicyclohexylammonium salt, boc-n-me-phe-oh dicyclohexylammonium salt chn, n-alpha-t-butyloxycarbonyl-n-alpha-methyl-l-phenylalanine dicyclohexylamine
CC(C)(C)OC(=O)N(C)C(CC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2
460.659
460.65
97%
25g
40163-88-0
MFCD00070397
MFORGORUOHHLHW-YDALLXLXSA-N
N-cyclohexylcyclohexanamine;(2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-phenylpropanoic acid
16211321
Solid
174°C to 177°C
Videos
SDS Documentos