Learn More
Alfa Aesar™ N-Boc-L-phenylalanine N-succinimidyl ester, 98%
Marca:  Alfa Aesar™ H62353.03
Detalles adicionales : CAS : 3674-06-4 Peso : 0.00100kg
Cantidad. 1g
 Ver más versiones de este producto
Código de producto. 15448878
  • 83.30€ / 1 g
Fecha de envío estimada
del proveedor 23-07-2019
Añadir a la cesta
Especificaciones
3674-06-4
362.38
NHUCANAMPJGMQL-ZDUSSCGKSA-N
107447
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
C18H22N2O6
MFCD00037911
boc-phe-osu, boc-l-phenylalanine n-hydroxysuccinimide ester, tert-butyl s-1-benzyl-2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, 2,5-dioxopyrrolidin-1-yl 2s-2-tert-butoxycarbonyl amino-3-phenylpropanoate, ambotzbaa5880, boc-phe-osu, n-boc-phe-succinimidyl oh, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl-l-phenylalaninate, boc-phe-osu hplc, boc-l-phenylalanine n-hydroxy succinimide ester
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
Especificaciones
N-Boc-L-phenylalanine N-succinimidyl ester
C18H22N2O6
715573
NHUCANAMPJGMQL-ZDUSSCGKSA-N
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoate
107447
Powder
White
1g
3674-06-4
MFCD00037911
boc-phe-osu, boc-l-phenylalanine n-hydroxysuccinimide ester, tert-butyl s-1-benzyl-2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, 2,5-dioxopyrrolidin-1-yl 2s-2-tert-butoxycarbonyl amino-3-phenylpropanoate, ambotzbaa5880, boc-phe-osu, n-boc-phe-succinimidyl oh, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl-l-phenylalaninate, boc-phe-osu hplc, boc-l-phenylalanine n-hydroxy succinimide ester
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
362.38
362.38
98%
150°C to 152°C
Hygroscopic