missing translation for 'onlineSavingsMsg'
Learn More

Éster de 1-terc-butilo de ácido N-Boc-D-aspártico, 98 %, Thermo Scientific Chemicals

Catalog Number 15442809
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
77004-75-2
C13H23NO6
289.328
MFCD00236827
RAUQRYTYJIYLTF-MRVPVSSYSA-N
boc-d-asp-otbu, r-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-boc-d-aspartic acid 1-tert-butyl ester, r-4-tert-butoxy-3-boc-amino-4-oxobutanoic acid, 3r-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-d-aspotbu, n-boc-d-asparticacid1-tert-butyl ester, r-4-tert-butoxy-3-boc-amino-4-oxobutanoicacid, n-tert-butyloxycarbonyl-d-aspartic acid 1-tert-butyl ester
7019660
Ácido (3R)-4-[(2-metilpropan-2-il)oxi]-3-[(2-metilpropan-2-il)oxicarbonilamino]-4-oxobutanoico
CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C
Este artículo no se puede devolver. Vea la política de devoluciones
77004-75-2
C13H23NO6
289.328
MFCD00236827
RAUQRYTYJIYLTF-MRVPVSSYSA-N
boc-d-asp-otbu, r-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-boc-d-aspartic acid 1-tert-butyl ester, r-4-tert-butoxy-3-boc-amino-4-oxobutanoic acid, 3r-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-d-aspotbu, n-boc-d-asparticacid1-tert-butyl ester, r-4-tert-butoxy-3-boc-amino-4-oxobutanoicacid, n-tert-butyloxycarbonyl-d-aspartic acid 1-tert-butyl ester
7019660
Ácido (3R)-4-[(2-metilpropan-2-il)oxi]-3-[(2-metilpropan-2-il)oxicarbonilamino]-4-oxobutanoico
CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-Boc-D-aspartic acid 1-tert-butyl ester
Beilstein 3553120
Formula Weight (peso de la fórmula) 289.33
Porcentaje de pureza 98%
Cantidad 1 g
Forma física Sólido

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'