Learn More
N-Boc-2,4,5-trifluoro-D-fenilalanina, 98 %, Thermo Scientific™
Catalog Number 15439328
Click to view available options
Cantidad:
250 mg
1 g
5 g
486460-09-7
C14H16F3NO4
319.28
MFCD06659118
FTWHJNNFVISGNQ-LLVKDONJSA-N
r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-2,4,5-trifluoro-d-phenylalanine, 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-2,4,5-trifluorophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-d-2-amino-3-2,4,5-trifluoro-phenyl alanine, r-n-tert-butoxycarbonyl-2,4,5-trifluorophenylalanine, r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoicacid
7146200
Ácido (2R)-2-[(2-metilpropan-2-il)oxicarbonilamino]-3-(2,4,5-trifluorofenil)propanoico
CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1F)F)F)C(=O)O
486460-09-7
C14H16F3NO4
319.28
MFCD06659118
FTWHJNNFVISGNQ-LLVKDONJSA-N
r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-2,4,5-trifluoro-d-phenylalanine, 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-2,4,5-trifluorophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoic acid, boc-d-2-amino-3-2,4,5-trifluoro-phenyl alanine, r-n-tert-butoxycarbonyl-2,4,5-trifluorophenylalanine, r-2-tert-butoxycarbonyl amino-3-2,4,5-trifluorophenyl propanoicacid
7146200
Ácido (2R)-2-[(2-metilpropan-2-il)oxicarbonilamino]-3-(2,4,5-trifluorofenil)propanoico
CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1F)F)F)C(=O)O
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-Boc-2,4,5-trifluoro-D-phenylalanine
Formula Weight (peso de la fórmula) 319.28
Porcentaje de pureza 98%
Cantidad 1 g
Forma física Sólido