Learn More
N-Acetylsulfathiazole-d4 (Major), TRC
Product Code. 30326793
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30326793 50 mg 50mg
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30326793 Supplier Toronto Research Chemicals Supplier No. TRCA18797650MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Acetylsulfathiazole-d4
CAS 1020718-91-5
Molecular Formula C11 2H4 H7 N3 O3 S2
Quantity 50 mg
Synonym N-Acetylsulfathiazole D4
InChI Formula InChI=1S/C11H11N3O3S2/c1-8(15)13-9-2-4-10(5-3-9)19(16,17)14-11-12-6-7-18-11/h2-7H,1H3,(H,12,14)(H,13,15)/i2D,3D,4D,5D
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)C)S(=O)(=O)Nc2nccs2
IUPAC Name N-[2,3,5,6-tetradeuterio-4-(1,3-thiazol-2-ylsulfamoyl)phenyl]acetamide
Molecular Weight (g/mol) 301.38
Formula Weight 301.0493