missing translation for 'onlineSavingsMsg'
Learn More

Monohidrato de N-acetil-D-manosamina, 99 %, Thermo Scientific Chemicals

Catalog Number p-7076767
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
1071625-31-4
C8H17NO7
239.22
MFCD00149493,MFCD00046758,MFCD00149493,MFCD00069808
VVQPUTSNIMAJPT-UHFFFAOYNA-N
n-acetylmannosamine
11908605
N-[(2R,3S,4S,5S,6R)-2,4,5-trihidroxi-6-(hidroximetil)oxan-3-il]acetamida
O.CC(=O)NC1C(O)OC(CO)C(O)C1O
Este artículo no se puede devolver. Vea la política de devoluciones
1071625-31-4
C8H17NO7
239.22
MFCD00149493,MFCD00046758,MFCD00149493,MFCD00069808
VVQPUTSNIMAJPT-UHFFFAOYNA-N
n-acetylmannosamine
11908605
N-[(2R,3S,4S,5S,6R)-2,4,5-trihidroxi-6-(hidroximetil)oxan-3-il]acetamida
O.CC(=O)NC1C(O)OC(CO)C(O)C1O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-Acetyl-D-mannosamine monohydrate
Punto de fusión ∼130°C (decomposition)
Envase Frasco de vidrio
Cantidad 250 mg
Beilstein 1346524
Formula Weight (peso de la fórmula) 239.24 (221.22 anhydrous)
Porcentaje de pureza 99%
Forma física Polvo

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'