missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Etoxifenil)-3-metilbenzamida, 97 %, Thermo Scientific™

Catalog Number p-7039730
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
328023-30-9
C16H17NO2
255.317
MFCD00595497
MMLNIRUCJDCWDS-UHFFFAOYSA-N
n-4-ethoxyphenyl-3-methylbenzamide, n-4-ethoxy-phenyl-3-methyl-benzamide, n∼1∼-4-ethoxyphenyl-3-methylbenzamide, n-4-ethoxyphenyl 3-methylphenyl carboxamide
675834
N-(4-etoxifenil)-3-metilbenzamida
CCOC1=CC=C(C=C1)NC(=O)C2=CC(=CC=C2)C
Este artículo no se puede devolver. Vea la política de devoluciones
328023-30-9
C16H17NO2
255.317
MFCD00595497
MMLNIRUCJDCWDS-UHFFFAOYSA-N
n-4-ethoxyphenyl-3-methylbenzamide, n-4-ethoxy-phenyl-3-methyl-benzamide, n∼1∼-4-ethoxyphenyl-3-methylbenzamide, n-4-ethoxyphenyl 3-methylphenyl carboxamide
675834
N-(4-etoxifenil)-3-metilbenzamida
CCOC1=CC=C(C=C1)NC(=O)C2=CC(=CC=C2)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-(4-Ethoxyphenyl)-3-methylbenzamide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 255.32
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'