missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Clorobencil)-3,5-dinitrobenzamida, 97 %, Thermo Scientific™

Catalog Number p-7039476
Click to view available options
Cantidad:
100 mg
500 mg
Este artículo no se puede devolver. Vea la política de devoluciones
446855-41-0
C14H10ClN3O5
335.7
MFCD00757362
ZDFCVPAIESTUKH-UHFFFAOYSA-N
n-4-chlorobenzyl-3,5-dinitrobenzamide, n-4-chlorophenyl methyl-3,5-dinitrobenzamide, 3,5-dinitrophenyl-n-4-chlorophenyl methyl carboxamide
2755819
N-[(4-clorofenil)metil]-3,5-dinitrobenzamida
C1=CC(=CC=C1CNC(=O)C2=CC(=CC(=C2)[N+](=O)[O-])[N+](=O)[O-])Cl
Este artículo no se puede devolver. Vea la política de devoluciones
446855-41-0
C14H10ClN3O5
335.7
MFCD00757362
ZDFCVPAIESTUKH-UHFFFAOYSA-N
n-4-chlorobenzyl-3,5-dinitrobenzamide, n-4-chlorophenyl methyl-3,5-dinitrobenzamide, 3,5-dinitrophenyl-n-4-chlorophenyl methyl carboxamide
2755819
N-[(4-clorofenil)metil]-3,5-dinitrobenzamida
C1=CC(=CC=C1CNC(=O)C2=CC(=CC(=C2)[N+](=O)[O-])[N+](=O)[O-])Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-(4-Chlorobenzyl)-3,5-dinitrobenzamide
Cantidad 100 mg
Formula Weight (peso de la fórmula) 335.7
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'