missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromo-2-fluorofenil)-4-clorobenzamida, 97 %, Thermo Scientific™

Catalog Number p-7039936
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
519017-62-0
C13H8BrClFNO
328.57
MFCD01359597
XLPWNTBBCDMYQV-UHFFFAOYSA-N
n-4-bromo-2-fluorophenyl-4-chlorobenzamide
4343155
N-(4-bromo-2-fluorofenil)-4-clorobenzamida
FC1=C(NC(=O)C2=CC=C(Cl)C=C2)C=CC(Br)=C1
Este artículo no se puede devolver. Vea la política de devoluciones
519017-62-0
C13H8BrClFNO
328.57
MFCD01359597
XLPWNTBBCDMYQV-UHFFFAOYSA-N
n-4-bromo-2-fluorophenyl-4-chlorobenzamide
4343155
N-(4-bromo-2-fluorofenil)-4-clorobenzamida
FC1=C(NC(=O)C2=CC=C(Cl)C=C2)C=CC(Br)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-(4-Bromo-2-fluorophenyl)-4-chlorobenzamide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 328.57
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'