Learn More
N-(4-Aminobutyl)Acetamide, TRC
Product Code. 30326683
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30326683 500 mg 500mg
1 options
This item is not returnable. View return policy
Product Code. 30326683 Supplier Toronto Research Chemicals Supplier No. TRCA577033500MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(4-Aminobutyl)Acetamide
CAS 5699-41-2
Molecular Formula C6H14N2O
Quantity 500 mg
Synonym (4-(Acetylamino)butyl)amine, Acetylputrescine, Monoacetylputrescine, N-Acetylputrescine, N1-Acetyl putrescine
InChI Formula InChI=1S/C6H14N2O/c1-6(9)8-5-3-2-4-7/h2-5,7H2,1H3,(H,8,9)
SMILES CC(=O)NCCCCN
IUPAC Name N-(4-aminobutyl)acetamide
Molecular Weight (g/mol) 130.19
Formula Weight 130.1106