missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Cloro-4-metilfenil)-2-metoxibenzamida, 97 %, Thermo Scientific™

Catalog Number p-7040103
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
313667-46-8
C15H14ClNO2
275.732
MFCD00702336
SASNELOZVTWLSU-UHFFFAOYSA-N
n-3-chloro-4-methylphenyl-2-methoxybenzamide, cambridge id 5303656
2189574
N-(3-cloro-4-metilfenil)-2-metoxibenzamida
CC1=C(C=C(C=C1)NC(=O)C2=CC=CC=C2OC)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
313667-46-8
C15H14ClNO2
275.732
MFCD00702336
SASNELOZVTWLSU-UHFFFAOYSA-N
n-3-chloro-4-methylphenyl-2-methoxybenzamide, cambridge id 5303656
2189574
N-(3-cloro-4-metilfenil)-2-metoxibenzamida
CC1=C(C=C(C=C1)NC(=O)C2=CC=CC=C2OC)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-(3-Chloro-4-methylphenyl)-2-methoxybenzamide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 275.73
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'