Learn More
N-(2-Aminoetil)pirrolidina, 99 %, Thermo Scientific Chemicals
Catalog Number p-3735418
Click to view available options
Cantidad:
5 mL
25 mL
100 mL
Envase:
Frasco de vidrio
7154-73-6
C6H14N2
114.19
MFCD00003182
WRXNJTBODVGDRY-UHFFFAOYSA-N
1-2-aminoethyl pyrrolidine, 1-pyrrolidineethanamine, n-2-aminoethyl pyrrolidine, 2-pyrrolidin-1-yl ethanamine, pyrrolidinoethylamine, pyrrolidinoethanamine, 2-1-pyrrolidinyl ethylamine, 2-pyrrolidin-1-yl ethan-1-amine, 2-pyrrolidinoethyl amine, 2-pyrrolidinoethylamine
1344
2-pirrolidin-1-iletanamina
C1CCN(C1)CCN
7154-73-6
C6H14N2
114.19
MFCD00003182
WRXNJTBODVGDRY-UHFFFAOYSA-N
1-2-aminoethyl pyrrolidine, 1-pyrrolidineethanamine, n-2-aminoethyl pyrrolidine, 2-pyrrolidin-1-yl ethanamine, pyrrolidinoethylamine, pyrrolidinoethanamine, 2-1-pyrrolidinyl ethylamine, 2-pyrrolidin-1-yl ethan-1-amine, 2-pyrrolidinoethyl amine, 2-pyrrolidinoethylamine
1344
2-pirrolidin-1-iletanamina
C1CCN(C1)CCN
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-(2-Aminoethyl)pyrrolidine
% mín. CAS 98.5
% máx. CAS 100.0
Color Incoloro a marrón
Densidad 0.9000g/mL
Punto de ebullición 66°C to 70°C (23.0 mmHg)
Temperatura de inflamación 47°C
Espectro infrarrojo Auténtico
Intervalo de porcentaje de ensayo 99%
Envase Frasco de vidrio
Índice de refracción 1.4677 to 1.4697
Cantidad 5 mL
Gravedad específica 0.9
Formula Weight (peso de la fórmula) 114.19
Porcentaje de pureza 99%
Forma física Líquido