missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dibromofenil)-4-metoxibenzamida, 97 %, Thermo Scientific™

Catalog Number p-7039979
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
541545-55-5
C14H11Br2NO2
385.055
MFCD03367365
MEEOWNOOARIQDA-UHFFFAOYSA-N
n-2,4-dibromophenyl-4-methoxybenzamide
4692851
N-(2,4-dibromofenil)-4-metoxibenzamida
COC1=CC=C(C=C1)C(=O)NC2=C(C=C(C=C2)Br)Br
Este artículo no se puede devolver. Vea la política de devoluciones
541545-55-5
C14H11Br2NO2
385.055
MFCD03367365
MEEOWNOOARIQDA-UHFFFAOYSA-N
n-2,4-dibromophenyl-4-methoxybenzamide
4692851
N-(2,4-dibromofenil)-4-metoxibenzamida
COC1=CC=C(C=C1)C(=O)NC2=C(C=C(C=C2)Br)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-(2,4-Dibromophenyl)-4-methoxybenzamide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 385.05
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'