Learn More
N-(1-Deoxy-D-fructos-1-yl)-L-methionine, TRC
Specifications
Specifications
| Chemical Name or Material | N-(1-Deoxy-D-fructos-1-yl)-L-methionine |
| CAS | 20638-92-0 |
| Molecular Formula | C11 H21 N O7 S |
| Quantity | 25 mg |
| Synonym | (S)-4-(Methylthio)-2-(((3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexyl)amino)butanoic Acid |
| InChI Formula | InChI=1S/C11H21NO7S/c1-20-3-2-6(11(18)19)12-4-7(14)9(16)10(17)8(15)5-13/h6,8-10,12-13,15-17H,2-5H2,1H3,(H,18,19)/t6-,8+,9+,10+/m0/s1 |
| SMILES | CSCC[C@H](NCC(=O)[C@@H](O)[C@H](O)[C@H](O)CO)C(=O)O |
| IUPAC Name | (2S)-4-methylsulfanyl-2-[[(3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexyl]amino]butanoic acid |
| Molecular Weight (g/mol) | 311.352 |
| Formula Weight | 311.104 |