missing translation for 'onlineSavingsMsg'
Learn More

MYRIP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP2-87871

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214016

  • 455.71€ / 100 microlitros
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

MYRIP Polyclonal specifically detects MYRIP in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

MYRIP
Polyclonal
Unconjugated
PBS, 2% Sucrose
Rabbit
Affinity purified
RUO
25924
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
DKFZp586F1018, exophilin-8, FLJ44025, MGC130034, myosin VIIA and Rab interacting protein, Myosin-VIIa- and Rab-interacting protein, MyRIP, rab effector MyRIP, SLAC2-C, SLAC2CMGC130035, Slp homolog lacking C2 domains c, Slp homologue lacking C2 domains, synaptotagmin-like protein homologue lacking C2 domains-c
The immunogen is a synthetic peptide directed towards the middle region of human MYRIP
100 μL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales