Learn More
Gilson™ MyPIPETMAN™ Enterprise Multichannel Pipettes
The Enterprise pipettes combine reliability, ergonomics, and personalization options to ensure efficiency and answer your ranging pipetting needs.
Marca:  Gilson™ FP10012E
Detalles adicionales : Peso : 0.19400kg
 Ver más versiones de este producto
Código de producto. 17793178
  • 1095.00€ / cada uno
Fecha estimada de envío: 08-06-2023
para ver el stock
Añadir a la cesta
Descripción
Descripción
MyPIPETMAN™ Enterprise pipettes are produced in a series of at least 150 units (in a mix of different volumes). All pipettes inside a series have the same technical features, design, and information printed on the handle.
  • Select your preferred technical features to optimize the use of your pipette and overall productivity in the lab
  • Color choices and printing options on your pipette allow for better organization and identification within your workflow
Especificaciones
Especificaciones
Diamond Tips, Amplipur Tips
Autoclavable
No
Polypropylene, PVDF, PBT
12
Non-sterile
20 μL
No
Assorted
Textured
P12x20
MyPipetman™ Enterprise
Mechanical Pipette