missing translation for 'onlineSavingsMsg'
Learn More

Myosin Light Chain 2 Antibody (3B2), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-30249FR

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215175

  • 521.58€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Myosin Light Chain 2 Monoclonal specifically detects Myosin Light Chain 2 in Human, Mouse, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

Myosin Light Chain 2
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
MYL2
Recombinant human MYL2 (1-166aa) purified from E. coli (NP_000423).
0.1 mL
Phospho Specific
4633
Human, Mouse, Xenopus species
Purified
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
3B2
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2
Mouse
Protein G purified
RUO
Primary
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human MYL2 protein.
Store at 4C in the dark.
IgG2b κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales