missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYLK2 (skMLCK) Recombinant Human Protein
MYLK2 (skMLCK) Recombinant Human Protein
Marca: Invitrogen PV3757
Detalles adicionales : Peso : 0.05250kg
Descripción
MYLK2 (skMLCK) is a calcium⁄calmodulin-dependent serine⁄threonine kinase expressed exclusively in adult skeletal muscle.Especificaciones
NP_149109 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
92.1kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
MYLK2 (skMLCK) | |
MYLK2 (skMLCK) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 10μg/mL Calmodulin, 0.5mM CaCl2, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 100μg/mL MLCK peptide substrate (KKRAARATSDVFA-NH2) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
MYLK2 (skMLCK) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.18mg/mL | |
114294, 85366 | |
MYLK2 (skMLCK) Recombinant Human Protein | |
–80°C | |
1 Each | |
MYLK2 (skMLCK) Recombinant Human Protein | |
MLCK | |
5870nmoles of phosphate transferred to MLCK peptide substrate (KKRAARATSDVFA-NH2) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.21μg/mL | |
GST Tag | |
Kinase and inhibitors | |
95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.