missing translation for 'onlineSavingsMsg'
Learn More

MYL3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2E4B2, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66286-1-IG-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818025

  • 109.00€ / 20 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy.
Especificaciones

Especificaciones

MYL3
2E4B2
Unconjugated
MYL3
Liquid
Myl3
MYL3 Fusion Protein Ag24535
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Porcine, Rat
Western Blot, Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08590, P09542, P16409
Cardiac myosin light chain 1, CMH8, CMLC1, MLC1SB, MLC1V, MYL3, Myosin light chain 3, VLC1
Mouse
IgG1
20 μL
-20°C
Primary
100515755, 17897, 24585, 4634
Videos
SDS
Documentos
Certificados