Learn More
Lonza™ MycoZap™ Plus-PR
Product Code. 11566529
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
11566529 10 x 1 mL Pack of 10
1 options
This item is not returnable. View return policy
Product Code. 11566529 Supplier Lonza™ Supplier No. VZA2021
  • ONLINE EXCLUSIVE PRICE
    391.00€
    305.86€ / paquete de 10
    Ahorro 85.14€
    21% Descuento
 Restricted Item
Fecha estimada de envoi: 24-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Antibiotic formulation for prevention of bacteria, fungi, and mycoplasma contaminations in primary cell cultures.
  • Active against mycoplasma, Gram negative and Gram positive bacteria, as well as yeast and fungi.
  • Complete solution replacing Pen/Strep formulation.
  • Applications:
    • Optimized for gentle protection of primary cells.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Prevention of cell culture contamination
pH 4.5 at 20°C
Quantity 10 x 1 mL