Learn More
Fisherbrand™ multiSUB™ Midi Horizontal Gel System
Product Code. 12307287 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Sample Size:
50
100
Gel Dimensions:
W x L: 10 x 10 cm
W x L: 10 x 7 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Size Gel Dimensions Unit Size
12307287 50 W x L: 10 x 7 cm Each
12397277 100 W x L: 10 x 10 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12307287
  • ONLINE EXCLUSIVE PRICE
    556.00€
    372.56€ / paquete de 1
    Ahorro 183.44€
    32% Descuento
View all promotions
Fecha estimada de envoi: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Designed for routine horizontal gel electrophoresis.
  • Suitable for rapid electrophoresis
  • Low buffer volumes
  • Runs about 50 to 100 samples
Specifications
Type Horizontal Gel Electrophoresis System
Gel Dimensions W x L: 10 x 7 cm
Includes multiSUB™ Midi electrophoresis unit, 10 x 7cm UV Tray, 2 x 16 sample combs, loading guides and dams
Sample Size 50
Volume (English) Buffer 0.07 gal.
Volume (Metric) Buffer 300 mL
Length (English) 4.92 in.
Length (Metric) 12.5 cm
Width (English) 8.66 in.
Width (Metric) 22 cm
Height (English) 3.54 in.
Height (Metric) 9 cm