missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUCL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558653
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: AQNPTTAAPA DTYPATGPAD DEAPDAETTA AATTA Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 42%.
MUCL1 Polyclonal antibody specifically detects MUCL1 in Human samples. It is validated for Immunohistochemistry.Especificaciones
MUCL1 | |
Polyclonal | |
Unconjugated | |
MUCL1 | |
mucin like 1; mucin-like 1; mucin-like protein 1; MUCL1; Protein BS106; SBEM; Small breast epithelial mucin; UNQ590/PRO1160 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
118430 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.25 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DR8 | |
MUCL1 | |
Recombinant protein corresponding to Human MUCL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only