Learn More
Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1 Mouse Monoclonal Antibody, Epredia™
Deliver accurate, reproducible results with the Epredia™ Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-145-RQ
Detalles adicionales : Peso : 0.02000kg
Descripción
Mucins are high molecular eight glycoproteins with 80% carbohydrate contents and the remaining 20% is constituted by protein core. Gastric mucin is found in normal human gastrointestinal tract. Resurgence of gastric mucin reactivity during colonic carcinogenesis is due to re-expression of the peptide core of gastric (or fetal colonic) mucins.Host Species: Mouse
Clone: 45M1
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Rabbit, Cat, Mouse, Rat, Pig, Hedgehog and Chicken. Does not react with cow. Others not known.
Epitope: Destroyed by β-mercaptoethanol and proteases but not by periodate treatment
Immunogen: M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient
Molecular Weight: >1000kDa
Positive Control: Stomach
Cellular Localization: Cytoplasmic and cell surface
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1 | |
Monoclonal | |
Unconjugated | |
M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient | |
IVD | |
Primary |
Immunohistochemistry (Paraffin) | |
45M1 | |
Mouse | |
12 mL | |
Cancer and Tumor Biology | |
Ready-to-Use |