missing translation for 'onlineSavingsMsg'
Learn More

MSRA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 14547-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16813244

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Methionine sulfoxide reductase (MsrA) reduces methionine sulfoxide (MetO) residues in proteins and free MetO to Methionine (Met). The catalytic activity of MsrA is dependent of bound metal and cofactors but it requires reducing equivalents from either DTT or a thioredoxin-regenerating system. MsrA plays an essential role in protecting cells against oxidative damage. The substrates of MsrA include calmodulin, HIV protease and a 1-proteinase-inhibitor. Recent studies indicate that there is a connection between MsrA and Alzheimer′s disease in mammals.
Especificaciones

Especificaciones

MSRA
Polyclonal
Unconjugated
MSRA
MSRA, Peptide Met(O) reductase, PMSR
Rabbit
Antigen Affinity Chromatography
RUO
110265, 29447, 4482
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q923M1, Q9D6Y7, Q9UJ68
MSRA
MSRA Fusion Protein Ag6053
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales