missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSLN Mouse anti-Human, Clone: 2E3A4, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66404-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein that is present on the surface of normal mesothelium and is overexpressed in epithelial ovarian cancer and maligt mesotheliomas.Especificaciones
MSLN | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13421 | |
MSLN | |
MSLN Fusion Protein Ag23606 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2E3A4 | |
Unconjugated | |
MSLN | |
CAK1, CAK1 antigen, mesothelin, MPF, MPF Mesothelin, cleaved form, MSLN, SMR | |
Mouse | |
Protein G | |
RUO | |
10232 | |
-20°C | |
Liquid |