Learn More
MS4A12 Monoclonal Antibody (4F4), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA524481
Detalles adicionales : Peso : 0.01000kg
Descripción
Peptide Sequence: MMSSKPTSHA EVNETIPNPY PPSSFMAPGF QQPLGSINLE NQAQGAQRAQ PYGITSPGIF ASSQPGQGNI QMINPSVGTA VMNFKEEAKA LGVIQIMVGL MHIGFGIVLC LISFSFREVL GFASTAVIGG YPFWGGLSFI ISGSLSVSAS KELSRCLVKG SLGMNIVSSI LAFIGVILLL VDMCINGVAG QDYWAVLSGK GISATLMIFS LLEFFVACAT AHFANQANTT TNMSVLVIPN MYESNPVTPA SSSAPPRCNN YSANAPK
MS4A12 Monoclonal antibody specifically detects MS4A12 in Human samples. It is validated for ELISA, Western Blot.Especificaciones
MS4A12 | |
Monoclonal | |
Unconjugated | |
MS4A12 | |
membrane spanning 4-domains A12; membrane-spanning 4-domains subfamily A member 12; membrane-spanning 4-domains, subfamily A, member 12; Ms4a10; MS4A12 | |
Mouse | |
Protein A | |
RUO | |
54860 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
4F4 | |
PBS with no preservative; pH 7.4 | |
Q9NXJ0 | |
MS4A12 | |
MS4A12 (AAH29793, 1 a.a. ∽ 267 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |