missing translation for 'onlineSavingsMsg'
Learn More

MRVI1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA552592

Detalles adicionales : Peso : 0.02000kg

Código de producto. 15760541

  • 410.00€ / 100 microlitros


Fecha de envío estimada
del proveedor 11-06-2020
Añadir a la cesta

Descripción

Descripción

This gene is similar to a putative mouse tumor suppressor gene (Mrvi1) that is frequently disrupted by mouse AIDS-related virus (MRV). The encoded protein, which is found in the membrane of the endoplasmic reticulum, is similar to Jaw1, a lymphoid-restricted protein whose expression is downregulated during lymphoid differentiation. Studies in mouse suggest that MRV integration at Mrvi1 induces myeloid leukemia by altering the expression of a gene important for myeloid cell growth and/or differentiation, and thus this gene may function as a myeloid leukemia tumor suppressor gene. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene, few of which initiate translation at a non-AUG (CUG) start site.
Especificaciones

Especificaciones

MRVI1
Polyclonal
Unconjugated
MRVI1
Liquid
MRVI1
Recombit protein corresponding to Human MRVI1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9Y6F6
IRAG, JAW1L, IP3R-associated cGMP kinase substrate, JAW1-related protein MRVI1, inositol 1,4,5-triphosphate receptor-associated cGMP kinase substrate, protein MRVI1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10335
Videos
Documentos

Research Use Only