missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRRF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12357-2-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Responsible for the release of ribosomes from messenger RNA at the termination of protein biosynthesis. May increase the efficiency of translation by recycling ribosomes from one round of translation to another By similarity. Ota T., Nat. Genet. 36:40-45(2004). Humphray S.J., Nature 429:369-374(2004). Zhang Y., Biochim. Biophys. Acta 1443:245-250(1998).Especificaciones
MRRF | |
Polyclonal | |
Unconjugated | |
MRRF | |
MRFF, MRRF, MTRRF, RRF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
311903, 67871, 92399 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5RKI9, Q96E11, Q9D6S7 | |
Mrrf | |
MRRF Fusion Protein Ag3021 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |