Learn More
MRPS9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561750
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: GIDYQLYFPI TQDREQLMFP FHFVDRLGKH DVTCTVSGGG RSAQAGAIRL AMAKALCSFV TEDEVEWMRQ AGLLTTDPRV RERKKPGQEG ARRKFTWKK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
MRPS9 Polyclonal antibody specifically detects MRPS9 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
MRPS9 | |
Polyclonal | |
Unconjugated | |
MRPS9 | |
2310002A08Rik; 28S ribosomal protein S9, mitochondrial; AW322516; mg637; mitochondrial ribosomal protein S9; Mitochondrial small ribosomal subunit protein uS9m; MRPS9; MRP-S9; muscle protein 637; RPMS9; S9mt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64965 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P82933 | |
MRPS9 | |
Recombinant protein corresponding to Human MRPS9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only