missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA550061
Detalles adicionales : Peso : 0.01000kg
Descripción
The antibody detects endogenous levels of total MRPS5 protein.
MRPS5 Polyclonal antibody specifically detects MRPS5 in Human samples. It is validated for Immunohistochemistry (Paraffin).Especificaciones
MRPS5 | |
Polyclonal | |
Unconjugated | |
MRPS5 | |
1620401I16Rik; 28S ribosomal protein S5, mitochondrial; AI850294; mitochondrial 28S ribosomal protein S5; mitochondrial ribosomal protein S5; Mitochondrial small ribosomal subunit protein uS5m; MRPS5; MRP-S5; S5mt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64969 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P82675 | |
MRPS5 | |
Synthesized peptide derived from internal of human MRPS5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |