Learn More
MRPS25 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA560386
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: LREEEEEKKQ LSHPANFGPR KYCLRECICE VEGQVPCPSL VPLPKEMRGK YKAALKADAQ D Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
MRPS25 Polyclonal antibody specifically detects MRPS25 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
MRPS25 | |
Polyclonal | |
Unconjugated | |
MRPS25 | |
2810429N01Rik; 28S ribosomal protein S25, mitochondrial; mitochondrial 28S ribosomal protein S25; mitochondrial ribosomal protein S25; Mitochondrial small ribosomal subunit protein mS25; MRPS25; MRP-S25; RPMS25; rt25; S25mt; Shd10; Unknown (protein for MGC:134542) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64432 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P82663 | |
MRPS25 | |
Recombinant protein corresponding to Human MRPS25 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only