missing translation for 'onlineSavingsMsg'
Learn More

MRPS18A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

140.00€ - 399.00€

Especificaciones

Antígeno MRPS18A
Concentración 0.13 mg/mL
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16815804
Ver documentos Detalles de la promoción
Proteintech
16235-1-AP-20UL
20 μL
140.00€
20 microlitros
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 17-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16805804
Ver documentos Detalles de la promoción
Proteintech
16235-1-AP-150UL
150 μL
399.00€
150 microlitros
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 17-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S18P family. The encoded protein is one of three that has significant sequence similarity to bacterial S18 proteins. The primary sequences of the three human mitochondrial S18 proteins are no more closely related to each other than they are to the prokaryotic S18 proteins. A pseudogene corresponding to this gene is found on chromosome 3p. Alternative splicing results in multiple transcript variants.
Especificaciones

Especificaciones

MRPS18A
Western Blot
Unconjugated
Rabbit
Rat, Mouse, Human
Q99N85, Q9NVS2
301249, 55168, 68565
MRPS18A Fusion Protein Ag8981
Primary
-20°C
MRPS18A
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
HumanS18b, MRP S18 3, MRP S18 a, MRPS18 3, MRPS18A, S18bmt, S18mt a
MRPS18A
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documentos

Documentos

Product Certifications