missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL54 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17683-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.Especificaciones
MRPL54 | |
Polyclonal | |
Unconjugated | |
MRPL54 | |
L54mt, MRP L54, MRPL54 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116541, 66047 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6P161, Q9CPW3 | |
Mrpl54 | |
MRPL54 Fusion Protein Ag11782 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |