missing translation for 'onlineSavingsMsg'
Learn More

MRPL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA548834

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15603135

  • 339.00€ / 400 microlitros


Fecha de envío estimada
del proveedor 07-11-2019
Añadir a la cesta

Descripción

Descripción

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the EcoL2 ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 12q. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq, Jul 2014]
Especificaciones

Especificaciones

MRPL2
Polyclonal
PBS with 0.09% sodium azide
Q5T653
39S ribosomal protein L2, mitochondrial, L2mt, MRP-L2, MRPL2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51069
Western Blot
Unconjugated
MRPL2
Liquid
MRPL2
KLH conjugated synthetic peptide between 236-264 amino acids from the C-terminal region of human MRPL2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos
Documentos