missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA549939
Detalles adicionales : Peso : 0.01000kg
Descripción
The antibody detects endogenous levels of total MRPL15 protein.
MRPL15 Polyclonal antibody specifically detects MRPL15 in Human, Mouse samples. It is validated for Western Blot.Especificaciones
MRPL15 | |
Polyclonal | |
Unconjugated | |
Mrpl15 | |
39S ribosomal protein L15, mitochondrial; HSPC145; L15mt; Mitochondrial large ribosomal subunit protein uL15m; mitochondrial ribosomal protein L15; Mrpl15; MRP-L15; MRP-L7; ribosomal protein, mitochondrial, L7; RPML7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27395, 29088 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9CPR5, Q9P015 | |
Mrpl15 | |
Synthesized peptide derived from internal of human MRPL15. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |