missing translation for 'onlineSavingsMsg'
Learn More

MROH2A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA562553

Detalles adicionales : Peso : 0.02000kg

Código de producto. 15700352

  • 410.00€ / 100 microlitros


Fecha de envío estimada
del proveedor 15-06-2020
Añadir a la cesta

Especificaciones

Especificaciones

MROH2A
Polyclonal
Unconjugated
MROH2A
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
339766
Immunohistochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
A6NES4
MROH2A
Recombit protein corresponding to Human MROH2A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos
Documentos

Research Use Only