missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16392-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
MRM1 belongs to the RNA methyltransferase trmH family and probably methylates the ribose of guanosine G-2270 in the peptidyl transferase center of the mitochondrial large ribosomal RNA (21S).Especificaciones
MRM1 | |
Polyclonal | |
Unconjugated | |
MRM1 | |
MRM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217038, 363661, 79922 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6IN84, Q99J25 | |
Mrm1 | |
MRM1 Fusion Protein Ag9483 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |