missing translation for 'onlineSavingsMsg'
Learn More

MR1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA548846

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15623145

  • 339.00€ / 400 microlitros


Fecha de envío estimada
del proveedor 17-06-2020
Añadir a la cesta

Descripción

Descripción

Antigen-presenting molecule specialized in presenting microbial vitamin B metabolites. Involved in the development and expansion of a small population of T-cells expressing an invariant T-cell receptor alpha chain called mucosal-associated invariant T- cells (MAIT). MAIT lymphocytes are preferentially located in the gut lamina propria and therefore may be involved in monitoring commensal flora or serve as a distress signal. Expression and MAIT cell recognition seem to be ligand-dependent.
Especificaciones

Especificaciones

MR1
Polyclonal
PBS with 0.09% sodium azide
Q95460
Major histocompatibility complex class I-related gene protein, MHC class I-related gene protein, Class I histocompatibility antigen-like protein, MR1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3140
Western Blot
Unconjugated
MR1
Liquid
MR1
KLH conjugated synthetic peptide between 312-341 amino acids from the C-terminal region of human MR1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos
Documentos