Learn More
MP Biomedicals™ Peptone, Bacteriological Grade
A water-soluble mixture of polypeptides and amino acids formed by the partial hydrolysis of protein
Marca: MP Biomedicals™ 0210480801
Detalles adicionales : Peso : 0.45400kg
Descripción
Peptone is produced from porcine and/or bovine tissues. The tissues are digested enzymatically to create small peptides and then spray dried. It is used in culture media in bacteriology. The growth requirements of most non-fastidious organisms will be fulfilled by its range of amino acids, peptides and proteoses.
- Presentation: Tan powder
- Suitable for bacteriological growth media
- pH: 5.5 - 7.0 (10% aq soln)(Lit.)
- Sulphated Ash: Ash=≤4%
- Moisture Content: ≤10%
- Solubility: Water soluble (100 mg/mL - clear to slightly hazy solution)
Especificaciones
Microbial Culture | |
5.5 to 7 | |
Porcine and/or Bovine Tissues | |
Powder |
Peptone from Porcine and/or Bovine tissues | |
0.45kg (1 lb.) | |
15° to 30°C |