Learn More
MP Biomedicals™ Gentamicin Sulfate
Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis
Marca:  MP Biomedicals™ 0219453001
Detalles adicionales : CAS : 1405-41-0 Peso : 0.00100kg
 Ver más versiones de este producto
Código de producto. 11490772
  • 245.00€ / 1 g
Fecha estimada de envío
07-06-2019
Añadir a la cesta
Descripción
Descripción
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
General antimicrobial effective against gram positive bacteria, gram negative bacteria, and mycoplasma.
Especificaciones
Especificaciones
Non-sterile
3.5 to 5.5
Gentamicin Sulfate
4°C
Powder
1g
4°C
Videos