Learn More
MP Biomedicals™ Bovine Collagen, Sterile
Aqueous solution
Marca: MP Biomedicals™ 0219349220
Detalles adicionales : CAS : 9007-34-5 Peso : 0.02000kg
Descripción
Contains 0.3% pure bovine collagen buffered with 0.1% acetate. 95% type I collagen, 5% type III collagen. pH (20 °C): 3.2-3.8.
Especificaciones
Contains 0.3% pure Bovine Collagen Buffered with 0.1% Acetate; 95% Type I Collagen, 5% Type III Collagen | |
1.7 to 2.1 | |
Bovine | |
Solution |
Collagen from Bovine | |
20mL | |
4°C |