missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse TNF-alpha Recombinant Protein
Mouse TNF-alpha Recombinant Protein
Marca: Invitrogen™ RMTNFAI
Detalles adicionales : Peso : 0.80000kg
Descripción
Mouse TNF-alpha Recombinant Protein for Western Blot, IP, ELISA, Ctrl
TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer. The mature, soluble form of mouse TNF-alpha was expressed in Pichia pastoris, KM71H. GenBank Accession &Num; P06804, amino acids 80 - 235. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: LRSSSQNSSD. Purity is >95% by SDS-PAGE analyses. Activity: The biological activity of recombinant mouse TNFa was measured by its cytolytic effect on mouse WEHI-13VAR cells in the presence of Actinomycin D. Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 0.5 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.Especificaciones
P06804 | |
PBS with no preservative | |
Mouse TNF alpha | |
Yeast | |
RUO | |
Mouse TNF alpha | |
Mouse | |
Ms | |
Recombinant | |
Recombinant |
Control, ELISA, Immunoprecipitation, Western Blot | |
21926 | |
20μg | |
4°C | |
TNF alpha; TNF-a; cachectin; tumor necrosis factor alpha; tumor necrosis factor ligand superfamily member 2; tumor necrosis factor-alpha | |
Tnf | |
Unconjugated | |
Yeast | |
Lyophilized | |
purified |