Learn More
Novus Biologicals™ Mouse Smad7 ELISA Kit (Colorimetric)
Mouse Smad7 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marca:  Novus Biologicals™ NBP2-76627
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213979
  • 780.76€ / 1 set
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
The Mouse Smad7 ELISA Kit (Colorimetric) measures mouse Smad7 in serum, plasma and other biological fluids.
Especificaciones
Especificaciones
0.313 to 20 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Mouse Smad7/MADH7 in samples. No significant cross-reactivity or interference between Mouse Smad7/MADH7 and analogues was observed.
SMAD7
1 Kit
0.188 ng/mL
Biological Fluids, Plasma, Serum
4092
Mouse
Storage of components varies. See protocol for specific instructions.