Learn More
STEMCELL Technologies Mouse Recombinant IL-22
Interleukin 22
Marca:  STEMCELL Technologies 78039
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17189411
  • 1431.08€ / 100 microgramos
Fecha estimada de envío: 14-11-2022
para ver el stock
Descripción
Descripción
  • Interleukin 22 (IL-22) is a class 2 α-helical cytokine that signals through the class 2 cytokine receptor and activates the JAK/STAT pathway
  • IL-22 is secreted by Th1, Th2, Tc22, and γδ T cells, dendritic, mast and NK cells
  • It stimulates expression of antimicrobial peptides from epithelial cells, thus hindering bacterial infections
  • Depending on the interactions with other cytokines, IL-22 can either promote inflammation or prevent tissue destruction by regulating host defense and tissue homeostasis at barrier surfaces (Sonnenberg et al.)
Especificaciones
Especificaciones
IL-TIF, Interleukin-22
≥95%
100 μg
Airway Cells; Hepatic Cells; Intestinal Cells; Keratinocytes; T Cells; T Cells, CD4+; T Cells, CD8+