Learn More
ACROBiosystems Mouse Flt-3 Ligand Protein, His Tag
423.38€ - 3108.82€
Especificaciones
Número de acceso | NP_038548.3 |
---|---|
Peso molecular | 20.2 kDa |
Fuente | Mouse Flt-3 Ligand, His Tag (FLL-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Arg 188 (Accession # P49772-1). |
Inmunógeno | Gly 27 - Arg 188 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16467549
|
ACROBiosystems
FLL-M52H4-100UG |
100 μg |
423.38€
100 microgramos |
Fecha estimada de envío: 02-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16477549
|
ACROBiosystems
FLL-M52H4-1MG |
1 mg |
3108.82€
1 miligramo |
Fecha estimada de envío: 02-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
FMS-like tyrosine kinase 3 ligand (Flt-3 Ligand) is also known as FL, Flt3L and FLT3LG, is an α-helical cytokine that promotes the differentiation of multiple hematopoietic cell lineages. FLT3LG is expressed as a noncovalentlylinked dimer by T cells and bone marrow and thymic fibroblasts. Each 36 kDa chain carries approximately 12 kDa of N- and O- linked carbohydrates. FLT3LG is structurally homologous to stem cell factor (SCF) and colony stimulating facor 1 (CSF-1). FLT3LG acts as a growth factor that increases the number of immune cells by activating the hematopoietic progenitors. It also induces the mobilization of the hematopoietic progenitors and stem cells in vivo which may help the system to kill cancer cells. FLT3LG induces the expansion of monocytes and immature dendritic cells as well as early B cell lineage differentiation. FLT3LG cooperates with IL2, IL6, IL7, and IL15 to induce NK cell development and with IL3, IL7 and IL11 to induce terminal B cell maturation. Animal studies also show FLT3LG to reduce the severity of experimentally induced allergic inflammation. FLT3LG is crucial for steady-state pDC and cDC development. A lack of FLT3L results in low levels of DCs.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 25-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_038548.3 | |
Mouse Flt-3 Ligand, His Tag (FLL-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Arg 188 (Accession # P49772-1). | |
-20°C | |
Flt-3 Ligand | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |
20.2 kDa | |
Gly 27 - Arg 188 | |
FLT3LG,FL,FLT3L,Flt3 ligand | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |