missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse E-Selectin/CD62E Biotinylated Antibody, R&D Systems™
Rat Monoclonal Antibody
Marca: R&D Systems BAM5752
552.50 EUR hasta el 2025-12-16
MEJOR PRECIO en promoción! Use el código promocional "24090" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
E-Selectin/CD62E Monoclonal specifically detects E-Selectin/CD62E in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Especificaciones
| E-Selectin/CD62E | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
| Q00690 | |
| SELE | |
| Mouse myeloma cell line NS0-derived recombinant mouse E-Selectin Trp22-Pro557 Accession # Q00690 | |
| 100 μg | |
| Primary | |
| Detects mouse E-Selectin/CD62E in ELISAs and Western blots. In Western blots, this antibody shows 20 - 50% cross-reactivity with recombinant human E-Selectin and recombinant mouse (rm) L-Selectin and no cross-reactivity with rmP-Selectin or recombinant rat E-Selectin. | |
| Mouse | |
| Purified |
| Western Blot, ELISA | |
| 96409 | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6401 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido