Learn More
Terminal Transferase (TdT) Mouse anti-Human, Brilliant Violet 421, Clone: E17-1519, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 565230
Detalles adicionales : Peso : 0.01000kg
Descripción
The E17-1519 monoclonal antibody specifically recognizes Terminal deoxynucleotidyl transferase (TdT), which is also known as Terminal transferase, or DNA nucleotidylexotransferase (DNTT). TdT is a 60 kDa nuclear polymerase that catalyzes the template-independent addition of deoxynucleotides to the 3'-hydroxyl termini of single-stranded oligonucleotide primer segments. TdT can contribute to antigen receptor diversity by N region diversification of rearranging immunoglobulin heavy chain and T cell receptor gene segments during pre-B and pre-T cell development. It is also expressed by certain lymphomas and leukemias.
The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Especificaciones
Terminal Transferase (TdT) | |
Monoclonal | |
Brilliant Violet 421 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Purified Human TdT | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry, Immunocytochemistry | |
E17-1519 | |
TdT; DNTT; Terminal transferase; Terminal addition enzyme | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |