missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rat IgG1/2a, Clone: G28-5, biotin, Secondary Antibody, BD

Marca:  BD Biosciences 553880

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15827448

  • 408.00€ / cada uno


Fecha estimada de envío
09-03-2020
Añadir a la cesta

Descripción

Descripción

The G28-5 antibody reacts specifically with rat IgG1 and IgG2a. It does not react with other Ig isotypes. A suspension of pooled rat IgG1 was used as the source of immunogen.

Host Species: Mouse
Clone: G28-5
Isotype: IgG2a
Species Reactivity: Rat
Immunogen: Pooled rat IgG

ELISA, Flow Cytometry

Especificaciones

Especificaciones

Mouse anti-Rat IgG1/2a
0.5mg/mL
Conjugated
IgG2a
RUO
Secondary
Rat
G28-5
Biotin
Murine
0.5mg
Immunology
Monoclonal
Videos
Documentos